Learn More
Medchemexpress LLC Sodium Tartrate | 868-18-8 | 99.0% | 25 MG
Supplier: Medchemexpress LLC 25MGHY128476
Sodium Tartrate is a pH-regulating agent with antioxidant activity. It is particularly effective in retarding hydrolysis when heated at high temperatures, which can increase the acid values (AVs) of vegetable oils.
- Acts as a pH-regulating agent
- Exhibits antioxidant activity
- Effective in retarding hydrolysis at high temperatures