Learn More
Medchemexpress LLC Sodium Tartrate | 868-18-8 | 99.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128476100MG
Encompass_Preferred
Sodium Tartrate is a pH-Regulating agent with antioxidant activity. It is particularly effective at retarding hydrolysis while heating at high temperatures, which increases the acid values (AVs) of vegetable oils.
  • Acts as a pH-regulating agent
  • Provides antioxidant activity
  • Retards hydrolysis during high-temperature heating
  • Increases acid values (AVs) of vegetable oils
Catalog No. 50-003-41830