Learn More
Strem, An Ascensus Company CAS# 26006-71-3. 25g. Sodium tellurate(VI) dihydrate, 99.5%. MFCD00150190
SDP
Supplier:  Strem, An Ascensus Company 93118625G
Encompass
CAS# 26006-71-3. 25g. Sodium tellurate(VI) dihydrate, 99.5%. MFCD00150190. Molecular Weight: 237.58 (273.61). Molecular Formula: Na2TeO4∙2H2O. Color/form: white pwdr. Strem# 93-1186. http://www.strem.com/catalog/v/93-1186/
Catalog No. 50-901-13909