Learn More
Strem, An Ascensus Company CAS# 1330-43-4. 10g. Sodium tetraborate, anhydrous (99.998%-B) (50 ppm K) PURATREM. MFCD00081185
SDP
Supplier:  Strem, An Ascensus Company 93107210G
Encompass
CAS# 1330-43-4. 10g. Sodium tetraborate, anhydrous (99.998%-B) (50 ppm K) PURATREM. MFCD00081185. Molecular Weight: 201.22. Molecular Formula: Na2B4O7. Color/form: white pwdr. Strem# 93-1072. http://www.strem.com/catalog/v/93-1072/
Catalog No. 50-901-13822