Learn More
Medchemexpress LLC Sodium tetradecyl sulfate (Tergitol 4) | 139-88-8 | 99.5% | C14H29NaO4S | 10 ML
Supplier: Medchemexpress LLC HYA019910ML
Sodium tetradecyl sulfate (Tergitol 4) is a scleroembolic agent and apoptosis inducer. It is used for research on varicose veins and vascular malformation diseases.
- Induces endothelial activation and release of endothelial particles in HUVEC.
- Increases expression of activation markers CD54 and CD62e.
- Inhibits proliferation and induces apoptosis in mouse endothelial tumor (EOMA) cells.