Learn More
Matrix Scientific Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]-borate, 79060-88-1, MFCD00043323, 1g
Supplier: Matrix Scientific 0091571G
Molecular Formula C32H12BF24Na, Purity 97%, Molecular Weight 886.21, Melting Point 300-306°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.