Learn More
Medchemexpress LLC Sodium tetraphenylborate | 143-66-8 | MFCD00011494 | 99.6% | 342.22 g·mol^-1 | C24H20BNa | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0257845G
Encompass_Preferred
Sodium tetraphenylborate is a white crystalline precipitating agent used in analytical chemistry for gravimetric determinations and selective precipitation of cations. Provided as a high-purity solid, it is suitable for laboratory research and analytical procedures.
  • High purity suitable for analytical applications.
  • Used as a precipitating agent in gravimetric determinations.
  • Stable solid form for easy handling and storage.
  • Compatible with common solvent systems for precipitation reactions.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-99665