Learn More
Medchemexpress LLC Thiocyanic acid, sodium salt | 540-72-7 | 99.2% | 81.07 | 1 ML
Supplier: Medchemexpress LLC HY2311910MM/1ML
Sodium thiocyanate reduces plasma levels of the pro-inflammatory cytokine IL-6 and increases the anti-inflammatory cytokine IL-10 levels. It also significantly reduces ROS formation. In vitro, Sodium thiocyanate attenuates atherosclerotic plaque formation and improves endothelial regeneration in mice, as well as inflammatory cytokine levels in the plasma and ROS and chlorotyrosine formation in the vessel wall.
- Reduces plasma levels of pro-inflammatory cytokine IL-6.
- Increases anti-inflammatory cytokine IL-10 levels.
- Significantly reduces ROS formation.
- Attenuates atherosclerotic plaque formation.
- Improves endothelial regeneration.