Learn More
Supelco Inc Sodium Thioglycolate, 367-51-1, MFCD00043386, 25 G
Supplier:  Supelco Inc T063225G
Encompass
Linear Formula: HSCH2COONa, Molecular Weight: 114.10, Synonym: Mercaptoacetic acid sodium salt, Thioglycolic acid sodium salt, ≥96.5% (iodometric), MP: >300 °C (lit.), storage temp: −20°C. Sodium thioglycolate has been used: as a growth supplement in enrichment media and to study its effect on Arcobacter, in influenza hemagglutinin formulation in order to reduce disulfide-mediated cross-linking and early potency loss, in electron microscopy. At room temperature, concentrations over 70% in water tend to form 1-2% thioglycolides per month which hydrolyze to the original free compound when made acidic or alkaline. The 70% solution oxidizes in air but is stable at room temperature when tightly closed. Thioglycolate salts may also lose purity on storage. The exclusion of air does not materially improve stability.
Catalog No. NC0734383