Learn More
Alkali Scientific Sodium Thiosulfate, 250 Grams
SDP
Supplier:  Alkali Scientific UMT002CP
Encompass_Preferred
Sodium Thiosulfate, laboratory grade, is available in a 250g quantity and is commonly used as a reducing agent in a variety of chemical reactions. This compound is particularly important in titration procedures, where it is used to neutralize iodine solutions in redox titrations. Sodium thiosulfate is also employed in photographic processing as a fixer, where it removes excess silver halide from exposed photographic film. In biochemical research, it is used in the preparation of solutions for sulfur studies and in the analysis of metals and other substances. Sodium thiosulfate is also utilized in laboratory protocols that involve dechlorination and decontamination.
Catalog No. 50-006-60603