Learn More
Alkali Scientific Sodium Thiosulfate, 5 Kilograms
SDP
Supplier:  Alkali Scientific UMT003CP
Encompass_Preferred
Sodium Thiosulfate, laboratory grade, is provided in a 5 kg weight and is widely used as a reagent in chemical analyses and industrial applications. It is commonly used in titrations, particularly in the determination of halogens like chlorine. Sodium thiosulfate acts as a reducing agent, converting iodine into iodide, which is key in redox reactions. It also serves as an essential component in photographic processing and the treatment of water for removing chlorine. In laboratory settings, sodium thiosulfate is crucial in various inorganic syntheses and analytical procedures requiring precise titrations and standardizations.
Catalog No. 50-006-60526