Learn More
Hach Company Sodium Thiosulfate, Pentahydrate, 454 g
Supplier:  Hach Company 46001
Encompass
ACS Reagent Grade, CAS No. 10102-17-7. Na2S2O3·5H2O, FW = 248.18. Assay: 99.5 - 101.0% Suitable for dechlorinating water samples and titrant preparation.
Catalog No. 50-199-4306