Learn More
Medchemexpress LLC Sodium thiosulfate pentahydrate | 10102-17-7 | MFCD00003499 | 99.99% | 248.18 | 100 G
SDP
Supplier:  Medchemexpress LLC HY23539C100G
Encompass_Preferred
Sodium thiosulfate pentahydrate, GR,≥99.5% is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It acts as an antioxidant and antifungal agent. It has been shown to protect against cisplatin-induced hearing loss in children and has demonstrated efficacy in the treatment of calcipylaxis and calcinosis cutis.
  • Can be used as a biological material or organic compound for life science related research.
  • Acts as an antioxidant and antifungal.
  • Protects against cisplatin-induced hearing loss.
  • May aid in the treatment of calcinosis cutis.
  • Effective and safe for the treatment of calcipylaxis.
Catalog No. 50-004-01437