Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium triphosphate pentabasic purum p.a., >=98.0% (T) | 7758-29-4 | MFCD00003514 | 500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 72061500G
Encompass
Sodium triphosphate pentabasic purum p.a., >=98.0% (T) | Purity: >=98.0% (T); 57-59% (calc. based on P2O5) | Mol Wt: 367.86 | 7758-29-4 | MFCD00003514 | 500G
Catalog No. 50-188-3712