Learn More
Selleck Chemical LLC Sofituzumab vedotin-D4036-1MG
SDP
Supplier:  Selleck Chemical LLC D40361MG
Encompass_Preferred
Sofituzumab Vedotin (DMUC5754A RG-7458) is an antibody-drug conjugate (ADC) that contains the humanized IgG1 anti-MUC16 monoclonal antibody (mAb) and a potent anti-mitotic agent monomethyl auristatin E (MMAE) linked through a protease-labile linker DMUC5754A binds to the MUC16 extracellular domain with high affinity
Catalog No. 50-313-4843