Learn More
Medchemexpress LLC Sofosbuvir impurity 1 | C₁₈H₁₇F₅NO₅P | 5 MG

Supplier: Medchemexpress LLC HY59121B5MG
Sofosbuvir impurity 1 is an impurity of Sofosbuvir, primarily intended for scientific research or drug declaration purposes. This product is not for personal use.
- Molecular weight: 453.30
- Appearance: Solid
- Color: White to off-white
- Shipping conditions: Room temperature in continental US (may vary elsewhere)
- Storage: Three years at room temperature; two years at -80°C in solvent, one year at -20°C in solvent
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.