Learn More
Medchemexpress LLC Sofosbuvir impurity A | 1496552-16-9 | 529.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15005C5MG
Encompass_Preferred
Sofosbuvir impurity A is a diastereoisomer of Sofosbuvir, which is an inhibitor of HCV RNA replication demonstrating potent anti-hepatitis C virus activity. This product is intended for research and analytical applications.
  • Diastereoisomer of Sofosbuvir
  • Inhibits HCV RNA replication
  • Potent anti-hepatitis C virus activity
  • Targeted for HCV
  • Solid appearance
  • White to yellow color
Catalog No. 50-003-59472