Learn More
Medchemexpress LLC Sofosbuvir impurity B | 96.20% | 529.45 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYI07195MG
Encompass_Preferred
Sofosbuvir impurity B is an impurity of Sofosbuvir. Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrating potent anti-hepatitis C virus (HCV) activity.
  • Active inhibitor of HCV RNA replication
  • Potent anti-hepatitis C virus (HCV) activity
  • Supplied as a white to off-white solid
Catalog No. 50-004-30770