Learn More
Medchemexpress LLC Sofosbuvir impurity F | 1337482-17-3 | 98.5% | 798.69 g/mol | C34H45FN4O13P2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYI040610MG
Encompass_Preferred
Sofosbuvir impurity F is a diastereoisomer and analytical reference impurity of the antiviral sofosbuvir, supplied as a light-yellow solid for research use only. It is intended for impurity profiling, analytical method development, and antiviral studies related to hepatitis C virus.
  • High purity reference at 98.5% for analytical standards.
  • Solid, light yellow to yellow appearance for easy handling.
  • Stable under recommended storage conditions as powder and in solvent.
  • Soluble in DMSO (100 mg/mL) for stock solution preparation.
  • Available in small mg pack sizes suitable for analytical testing.
Catalog No. 50-002-97706