Learn More
FUJIFILM Biosciences sOFTWARE
Catalog No. 5000450323
Encompass
Software that supports data processing based on the protocols complying with three types of pharmacopeia USP EP JP for bacterial endotoxin testing compliant with FDA 21 CFR Part 11 ERES electronic records/electronic signature and offers excellent audit trail capabilities can provide statistical processing of means standard deviations and has robust data processing functions The data processing functions can be performed by defining sample types such as standards controls and test samples
Catalog No. 50-004-50323 Supplier FUJIFILM Biosciences Supplier No. 29836871
Software that supports data processing based on the protocols complying with three types of pharmacopeia USP EP JP for bacterial endotoxin testing compliant with FDA 21 CFR Part 11 ERES electronic records/electronic signature and offers excellent audit trail capabilities can provide statistical processing of means standard deviations and has robust data processing functions The data processing functions can be performed by defining sample types such as standards controls and test samples