Learn More
LUNA NANOTECH INC Soil Genomic Dna Purification Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK06350
Encompass_Preferred
PuroSPIN™ Soil Genomic DNA Purification Kit, 50 Preps
The PuroSPIN™ Soil Genomic DNA Purification Kit  is designed to purify high molecular weigth genomic DNA from variety of microorganisms, fungi, bacteria, archaea, etc in soil samples. It greatly simplifies the purification process by incorporating a convenient and easy-to-use extraction technology based on nucleic acid binding to silica spin-columns which eliminates the use of toxic phenol-chloroform extraction or time consuming alcohol precipitation. The kit is suatable for different types of soil, such as: top-soil, bog, clay, forest, compost, etc.
Features:
- Easily purify genomic DNA from soil samples using silica spin columns
- Simple extraction process with no need for phenol or chloroform
- Lysate filtration step to remove any precipitated SDS-protein complexes, salts and other contaminants for higher yield and purity of genomic DNA products
Catalog No. 50-256-6951