Learn More
Medchemexpress LLC Solabegron | 252920-94-8 | MFCD11040855 | 99.9% | 410.89 | C23H23ClN2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1943610MG
Encompass_Preferred
Solabegron is a selective β3-adrenergic receptor agonist supplied for laboratory research use. It stimulates cAMP accumulation in cells expressing human β3-AR and has been investigated for treatment of overactive bladder and irritable bowel syndrome. The product is provided as a high-purity solid for in vitro and preclinical studies.
  • Selective β3-adrenergic receptor agonist.
  • Stimulates cAMP accumulation with reported EC50 ≈ 22 nM.
  • High purity (99.89%) suitable for analytical and biological assays.
  • Supplied as a solid in small-mass quantities for assay development.
  • Molecular weight 410.89; formula C23H23ClN2O3.
Catalog No. 50-002-94686