Learn More
Grainger Solar Fuse 100A PVS-R Series 600VDC
Supplier:  Grainger 14L661
Encompass_Preferred
The BUSSMANN Solar Fuse 100 Amp is a 100 amp solar fuse designed for photovoltaic systems. It has a voltage rating of 1000V DC and features a ceramic body construction with nickel-plated brass endcaps and blade terminals. This fast-acting Class T fuse provides overcurrent protection for PV source circuits, PV output circuits, and PV combiner boxes. It has a compact 13/32" x 1-1/2" body size. The BUSSMANN Solar Fuse line meets UL recognition requirements.
Catalog No. 50-275-2714