Learn More
Grainger Solar Fuse 70A PVS-R Series 600VDC
Supplier:  Grainger 14L658
Encompass_Preferred
The Bussmann 70 Amp Solar Fuse is a fast-acting, current-limiting fuse designed for photovoltaic system protection. It has a voltage rating of 1000V DC and interrupting rating of 200kA at 1000V DC. This Class J fuse features a ceramic body construction with nickel-plated brass endcaps and measures 1-1/4"L x 13/32"W. It can be used in PV combiner boxes and other DC equipment up to 70A. The compact design allows for high density panel designs.
Catalog No. 50-274-9836