Learn More
Grainger Solar Panel Controller 22V
Supplier:  Grainger 16W542
Encompass_Preferred
The Schumacher Electric Solar Panel Controller is an automatic solar charge controller that prevents overcharging and prolongs battery life. It is designed for 12V automotive and marine batteries. This pulse width modulation (PWM) controller provides up to 10 amps of charging current and works with solar panels up to 130 watts. It has LED indicators to show charging status and a temperature sensor to adjust charging voltage based on battery temperature. The controller has user adjustable settings for battery type, charging current, and load control. It is made with a waterproof enclosure and includes battery clamps and a wiring harness for easy installation.
Catalog No. 50-276-3661