Learn More
Selleck Chemical LLC Solasodine S3309-1mg
SDP
Supplier:  Selleck Chemical LLC S33091MG
Encompass_Preferred
Solasodine (Purapuridine Solancarpidine Solasodin Salasodine Salasdine) is a poisonous alkaloid chemical compound that occurs in plants of the Solanaceae family Solasodine reduces the mRNA level of matrix metalloproteinase-2 (MMP-2) MMP-9 and extracellular inducer of matrix metalloproteinase (EMMPRIN) but increases the expression of reversion-inducing cysteine-rich protein with kazal motifs (RECK) Solasodine downregulates oncogenic microRNA-21 (miR-21) which has been known to target RECK Solasodine also reduces PI3K/Akt signaling pathways and downregulates expression of miR-21
Catalog No. 50-310-9851