Learn More
TARGETMOL CHEMICALS INC SOLASONINE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T390110MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Solasonine exerts leishmanicidal activity against promastigote forms of L. amazonensis. purity: 99%
Catalog No. 50-257-6861