Learn More
TARGETMOL CHEMICALS INC Solcitinib 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T362010MG
Encompass_Preferred
Solcitinib (GSK2586184, GLPG0778) is a JAK1 inhibitor. Solcitinib may be potentially used for the treatment of psoriasis, ulcerative colitis, and systemic lupus erythematosus.
Catalog No. NC1623183