Learn More
Sparkfun Electronics SOLDER VACUUM
Supplier:  Sparkfun Electronics TOL13203
Encompass_Preferred
The Solder Vacuum, a great (and sometimes under appreciated) little tool for solder rework. It allows you to pull the molten solder out of a connection or via with ease. To use it you simply depress the plunger, heat up your solder, place the nozzle over the solder you want to remove and finally just push the button. The piston pops up and the solder disappears!
Catalog No. NC1071021