Learn More
Waters Corp V3 Solenoid
Supplier:  Waters Corp WAT025892
Encompass_Preferred
The V3 Solenoid Valve is a two-way, low pressure (1000psi maximum) rotary spool valve. This valve is used as V3, the Waste Valve, in the Fluid Pak of the 717 PLUS Autosampler.
Catalog No. 50-819-030