Learn More
Medchemexpress LLC (+)-Solketal | 22323-82-6 | 99.2% | 132.16 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01269910G
Encompass_Preferred
(+)-Solketal is a biochemical reagent designed for life science research, functioning as both a biological material and an organic compound.
  • Acts as a MEK inhibitor with antitumor activity.
  • Employed in the preparation of a chiral allylic triol through extrusion of sulfur dioxide from an alfa,beta-epoxysulfone.
Catalog No. 50-004-54813