Learn More
Cayman Chemical Solnum avcular Solasodn 10mg
SDP
Supplier:  Cayman Chemical 2811210 MG
Encompass_Preferred
An alkaloid with diverse biological activities; reduces glutamate-induced excitotoxicity in PC12 cells at 40 µM; inhibits cell invasion and migration of, as well as reverses TGF-β1-induced EMT in, HCT116 cells at 40 µM; reduces tumor growth and inhibits EMT in an HCT116 mouse xenograft model at 30 and 50 mg/kg; reduces carrageenan or arachidonic acid-induced paw edema at 75 mg/kg
Catalog No. 50-293-9452