Learn More
Agilent Technologies SOLOPACK GOLD SUPERCOMP CELLS
Supplier: Agilent Technologies 230350
SoloPack Gold Cells for Transformation in the Tube,convenient transformation in the tube, Now even easier, no BME required, robust growth produces large colonies.