Learn More
Sigma Aldrich Fine Chemicals Biosciences CyR3 Mono NHS Ester Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GEPA13101
Encompass
The packs contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups for example on antibodies peptides or oligonucleotides.
Catalog No. 50-290-4370