Learn More
Cayman Chemical Soluble BLySBAFFhumnrec 100ug
SDP
Supplier:  Cayman Chemical 31824100 UG
Encompass_Preferred
Source: Active recombinant N-terminal human IgG1 Fc-tagged BLyS/BAFF expressed in HEK293 cells • Amino acids: 134-285 • MW: 43.7 kDa
Catalog No. 50-295-0466