Learn More
Cayman Chemical Soluble TNFSF15humnrecom 100ug
SDP
Supplier:  Cayman Chemical 40855100 UG
Encompass_Preferred
Source: Recombinant human N-terminal His-tagged soluble TNFSF15 expressed in HEK293 cells • Amino Acids: 72-251 • Tag: N-terminal His • MW: 22.8 kDa
Catalog No. 50-294-5430