Learn More
PENTAIR AQUATIC ECO SYSTEMS SOLUTION CONDUCTIVITY 1000 S/C
Supplier:  PENTAIR AQUATIC ECO SYSTEMS Y3167E
Encompass
Solution Conductivity 1000 S/CM Per PintNIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy. 1 pint bottles.
Catalog No. NC2259272