Learn More
Ethos Biosciences Dichromate Cleaning Solution, 1gal, Ethos
Supplier:  Ethos Biosciences 3309G
Encompass_Preferred
Dichromate Cleaning Solution is a powerful oxidizing cleaning agent formulated for laboratory glassware and equipment. Commonly used to remove organic residues, grease, and stubborn contaminants, it provides effective, deep cleaning for flasks, beakers, pipettes, and other laboratory apparatus. The solution delivers consistent performance for routine maintenance and preparation of glassware used in analytical, chemical, and industrial procedures. Ideal for general laboratory cleaning, equipment decontamination, and preparation of surfaces prior to sensitive testing. Supplied in chemically compatible containers, it is suitable for research educational, and industrial laboratory environments.
Catalog No. 50-005-74311