Learn More
Cole-Parmer Cole-Parmer Cadmium (Cd+2) ISE Double junction solution kits
Supplier:  Cole-Parmer 2750257
Encompass_Preferred
These kits contain a 60 mL bottle of fill solution, 475 mL bottle of standard solution and 475 mL bottle of ISA.
Catalog No. 50-633-633
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov