Learn More
Selleck Chemical LLC Solutol HS-15-E1708-10G
SDP
Supplier:  Selleck Chemical LLC E170810G
Encompass_Preferred
Solutol HS-15(Polyethylene glycol 12-hydroxystearate) is a Macrogol 15 hydroxy stearate which improves permeability by disrupting the membrane structure/fluidity change Note Due to its chemical properties this product will be semi-solid and solid at room temperature and low temperature
Catalog No. 50-313-8583