Learn More
Chemglass Life Sciences Chemglass Solvent Reservoir System, 5 L, Complete (CG-1167-13)
SDP
Supplier:  Chemglass Life Sciences CG116713
Encompass_Preferred
Chemglass Solvent Reservoir System (5 L) is a complete plastic-coated glass reservoir kit designed to store and dispense HPLC mobile phases and other solvents. The system provides organized solvent storage, filtration, and quick-connect tubing for low-pressure liquid chromatography applications.
  • 5 L capacity plastic-coated glass reservoir.
  • Three-hole cap assembly compatible with GL45 threaded bottles.
  • Combination inlet/sparger filter with a 2 μm stainless steel filter cup.
  • Includes 10 ft of 1/8 in PTFE tubing, flangeless nuts, and ferrules for tubing connections.
  • Includes a Tefzel plug to seal unused ports.
Catalog No. NC2964714