Learn More
Metrohm USA SOLVOTRODE
SDP
Supplier:  Metrohm USA 60279300
Encompass
Combined pH glass electrode for titrations in nonaqueous media, with memory chip for sensor data such as article and serial number, calibration data and calibration history.
Catalog No. NC0567203