Learn More
Metrohm USA SOLVOTRODE EASYCLEAN
SDP
Supplier:  Metrohm USA 60229010
Encompass
The space-saving electrode for titration in nonaqueous media with easyClean diaphragm. Diaphragm: Ground-joint diaphragm; Electrode plug-in head: Fixed cable (l = 1 m) with plug F; Electrode slope: > 0.90; Electrode zero point (mV): 0-100 mV (pH = 7); Electrolyte flow rate (ul/h): 3-50; Indicator electrode shape: Sphere; Indicator electrode type: Glass electrode; Internal reference electrode type: LL system; Measuring range: 0-14; Measuring unit: pH; Membrane glass type: T; Membrane resistance: 50-150; Min. immersion depth (mm): 20; Plug for filling opening: Orifice; Reference electrolyte type: LiCl(sat) in ethanol; Reference system: LL system; Resistance (kOhm): <250; SGJ sleeve: flexible SGJ sleeve; Shaft diameter bottom (mm): 12; Shaft diameter top (mm): 12; Shaft material: Glass; Temperature range, long-term (C): 0-70; Temperature range, short-term (C): 0-70.
Catalog No. NC0367115