Learn More
Medchemexpress LLC Somatostatin-28 (sheep, human rat mouse) | 73032-94-7 | 99.7% | 3148.56 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP395410MG
Encompass_Preferred
Somatostatin-28 is a biologically active polypeptide, synthesized in the proximal intestinal epithelial cells. It suppresses glucose-stimulated insulin secretion without affecting circulating basal insulin concentration, and targets somatostatin receptor subtype 5 (SSTR5) to regulate GLP-1 secretion.
  • Biologically active polypeptide
  • Synthesized in proximal intestinal epithelial cells
  • Suppresses glucose-stimulated insulin secretion
  • Does not affect circulating basal insulin concentration
  • Targets somatostatin receptor subtype 5 (SSTR5)
  • Regulates GLP-1 secretion
Catalog No. 50-003-97246