Learn More
Selleck Chemical LLC Somatostatin Acetate P1039-1G
SDP
Supplier:  Selleck Chemical LLC P10391G
Encompass_Preferred
Somatostatin Acetate is a peptide hormone that regulates the endocrine system and affects neurotransmission and cell proliferation via interaction with G protein-coupled somatostatin receptors and inhibition of the release of numerous secondary hormones
Catalog No. 50-000-40822