Learn More
ABclonal Technology Somatostatin Receptor 2 (SSTR2) Rabbit pAb
SDP
Supplier:  ABclonal Technology A151011000UL
Encompass_Preferred
Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The biologic effects of somatostatin are probably mediated by a family of G protein-coupled receptors that are expressed in a tissue-specific manner. SSTR2 is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in cerebrum and kidney.
Catalog No. 50-001-07757