Learn More
Bioss Somatostatin Receptor 5 Monocl
SDP
Supplier:  Bioss BSM62315R
Encompass_Preferred
Somatostatin Receptor 5 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Rat Clonality Monoclonal Applications WB FCM RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-64929