Learn More
Medchemexpress LLC Sominone | 98569-64-3 | 458.63 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN108891MG
Encompass_Preferred
Sominone is the active metabolite of Withanoside IV. It enhances neuronal morphological plasticity by activating the RET pathway and induces axon/dendrite regeneration and synaptic reconstruction, which helps improve spatial memory. This compound is used in research for neurodegenerative diseases such as Alzheimer's disease.
  • Active metabolite of Withanoside IV.
  • Enhances neuronal morphological plasticity by activating the RET pathway.
  • Induces axon and dendrite regeneration and synaptic reconstruction.
  • Improves spatial memory.
  • Promotes axon and dendrite regeneration in β-amyloid treated rat cortical neurons.
  • Increases axonal and dendritic density in the hippocampus of ddY mice.
  • Shows improvement effects in AD model mice.
Catalog No. 50-004-14925