Learn More
Medchemexpress LLC Sominone | 98569-64-3 | 458.63 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN108895MG
Encompass_Preferred
Sominone is the active metabolite of Withanoside IV. It enhances neuronal morphological plasticity by activating the RET pathway, inducing axon/dendrite regeneration and synaptic reconstruction, thereby improving spatial memory. It can be used in the research of neurodegenerative diseases such as Alzheimer's disease.
  • Active metabolite of Withanoside IV.
  • Enhances neuronal morphological plasticity by activating the RET pathway.
  • Induces axon and dendrite regeneration and synaptic reconstruction.
  • Improves spatial memory.
  • Potential use in research for neurodegenerative diseases like Alzheimer's disease.
  • Enhances spatial memory and increases axonal and dendritic density in the hippocampus of ddY mice.
  • Shows improvement effects in AD model mice.
Catalog No. 50-004-16563