Learn More
Medchemexpress LLC Songorine | 509-24-0 | 357.49 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN208020MG
Encompass_Preferred
Songorine is a BBB-permeable diterpenoid alkaloid isolated from the genus *Aconitum*. It acts as a GABAA receptor antagonist in rat brain and possesses anti-cancer, antiarrhythmic, and anti-inflammatory activities. Songorine shows potential for the treatment of Epithelial Ovarian Cancer (EOC).
  • Bbb-permeable diterpenoid alkaloid
  • Isolated from the genus *Aconitum*
  • Gabaa receptor antagonist in rat brain
  • Anti-cancer activity
  • Antiarrhythmic activity
  • Anti-inflammatory activity
  • Potential for treatment of epithelial ovarian cancer (EOC)
Catalog No. 50-004-14794